Order Entry
Spec-Tec® Spectra® Fiber M102 Sterile Critical Environment Glove Liners
Spec-Tec® Spectra® Fiber M102 Sterile Critical Environment Glove Liners
Supplier:  WELLS LAMONT
Certificates
undefined
Spec-Tec® Spectra® Fiber M102 Sterile Critical Environment Glove Liners
Supplier:  WELLS LAMONT
Specifications
  • Catalog No:
  • 112-6613
  • 112-6610
  • 112-6611
  • 112-6612
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Cleanroom class:
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • Sterility:
  • Sterile
  • Sterile
  • Sterile
  • Sterile
Specifications
Product Information
  • Catalog #
    Supplier #
    Glove size
    Colour
    Length
    Pk
    Availability
    Price
  • 112-6612
    M102EU-S
    S
    White
    210 mm
    10 PAIR
    Log in for availability
    Log in for availability
    1 * 10 PÁR
    1 * 10 PÁR - 5 510,00 Kč
    5 510,00 Kč
    1 * 10 PÁR
       5 510,00 Kč
    Supplier Article Number: M102EU-S
    Local Article Number: WELLM102EU-S
    Specifications:
    More
    Specifications
    Ambidextrous / hand-specific
    Ambidextrous
    Environmentally Preferable
    Disposable / reusable
    Disposable
    Latex / latex-free
    Latex-free
    Powdered / powder-free
    Powder-free
    Cleanroom class
    10000/ISO 7
    Sterility
    Sterile
  • 112-6611
    M102EU-M
    M
    White
    210 mm
    10 PAIR
    Log in for availability
    Log in for availability
    1 * 10 PÁR
    1 * 10 PÁR - 5 510,00 Kč
    5 510,00 Kč
    1 * 10 PÁR
       5 510,00 Kč
    Supplier Article Number: M102EU-M
    Local Article Number: WELLM102EU-M
    Specifications:
    More
    Specifications
    Ambidextrous / hand-specific
    Ambidextrous
    Environmentally Preferable
    Disposable / reusable
    Disposable
    Latex / latex-free
    Latex-free
    Powdered / powder-free
    Powder-free
    Cleanroom class
    10000/ISO 7
    Sterility
    Sterile
  • 112-6610
    M102EU-L
    L
    White
    210 mm
    10 PAIR
    Log in for availability
    Log in for availability
    1 * 10 PÁR
    1 * 10 PÁR - 5 510,00 Kč
    5 510,00 Kč
    1 * 10 PÁR
       5 510,00 Kč
    Supplier Article Number: M102EU-L
    Local Article Number: WELLM102EU-L
    Specifications:
    More
    Specifications
    Ambidextrous / hand-specific
    Ambidextrous
    Environmentally Preferable
    Disposable / reusable
    Disposable
    Latex / latex-free
    Latex-free
    Powdered / powder-free
    Powder-free
    Cleanroom class
    10000/ISO 7
    Sterility
    Sterile
  • 112-6613
    M102EU-XL
    XL
    White
    210 mm
    10 PAIR
    Log in for availability
    Log in for availability
    1 * 10 PÁR
    1 * 10 PÁR - 5 510,00 Kč
    5 510,00 Kč
    1 * 10 PÁR
       5 510,00 Kč
    Supplier Article Number: M102EU-XL
    Local Article Number: WELLM102EU-XL
    Specifications:
    More
    Specifications
    Ambidextrous / hand-specific
    Ambidextrous
    Environmentally Preferable
    Disposable / reusable
    Disposable
    Latex / latex-free
    Latex-free
    Powdered / powder-free
    Powder-free
    Cleanroom class
    10000/ISO 7
    Sterility
    Sterile