(±)-2-Bromopalmitic acid
MOLEM18863611EA
0
CZK
MOLEM18863611
(±)-2-Bromopalmitic acid
(±)-2-Bromopalmitic acid
Vzorec:
H₃C(CH₂)₁₃BrCHCOOH MW: 335,32 g/mol Bod varu: 390 °C (1013 hPa) Bod tání: 53 °C |
CAS číslo:
18263-25-7 EINECS: 242-137-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...