3-Fluorostyrene, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIAL219452-1GEA
2160
CZK
SIAL219452-1G
3-Fluorostyrene, Sigma-Aldrich®
3-Fluorostyrene
Vzorec:
C₈H₇F MW: 122,14 g/mol Bod varu: 30 °C (1013 hPa) Hustota: 1,025 g/cm³ (20 °C) |
MDL Number:
MFCD00000341 CAS číslo: 350-51-6 EINECS: 206-504-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...