Antibacterial Tops, Long Sleeve, Base Pro
Supplier: Portwest
Certificates
About this item
The Base Pro top incorporates silver ion treatment, which combats odor and increases garment longevity.
- Provides insulation and thermal protection
- Unique anti-bacterial silver ion treatment
- Breathable, high-wicking, moisture management
- Designed with a comfort fit
- Contrast colour stitching for added style
- Retail box aids presentation for retail sales
The top is close fitting for optimal wicking performance and suitable for multi-season use. Recommended for dry daily wear. Black finish.
Specifications
- Catalog No:
- PRTWB153BKRXXL
- PRTWB153BKRXL
- PRTWB153BKRS
- PRTWB153BKRM
- PRTWB153BKRL
- Garment application:
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- Gender:
- For men
- For men
- For men
- For men
- For men
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Fabric:
- 93% wicking polyester, 7% elastane with silver ion treatment
- 93% wicking polyester, 7% elastane with silver ion treatment
- 93% wicking polyester, 7% elastane with silver ion treatment
- 93% wicking polyester, 7% elastane with silver ion treatment
- 93% wicking polyester, 7% elastane with silver ion treatment
- Weight:
- 0,284 g/m²
- 0,264 g/m²
- 0,22 g/m²
- 0,22 g/m²
- 0,242 g/m²
Specifications
Product Information
- Catalog #Supplier #ColourClothing SizeGarment fitPkAvailabilityPrice
- PRTWB153BKRSB153BKRSBlackSRegular11 * 1 KS763,00 KčSupplier Article Number: B153BKRSLocal Article Number: PRTWB153BKRS
Specifications
Garment applicationGeneral PurposeEnvironmentally PreferableRisk areaGenderFor menSleeve or leg lengthLongFabric93% wicking polyester, 7% elastane with silver ion treatmentWeight0,22 g/m² - PRTWB153BKRMB153BKRMBlackMRegular11 * 1 KS763,00 KčSupplier Article Number: B153BKRMLocal Article Number: PRTWB153BKRM
Specifications
Garment applicationGeneral PurposeEnvironmentally PreferableRisk areaGenderFor menSleeve or leg lengthLongFabric93% wicking polyester, 7% elastane with silver ion treatmentWeight0,22 g/m² - PRTWB153BKRLB153BKRLBlackLRegular11 * 1 KS763,00 KčSupplier Article Number: B153BKRLLocal Article Number: PRTWB153BKRL
Specifications
Garment applicationGeneral PurposeEnvironmentally PreferableRisk areaGenderFor menSleeve or leg lengthLongFabric93% wicking polyester, 7% elastane with silver ion treatmentWeight0,242 g/m² - PRTWB153BKRXLB153BKRXLBlackXLRegular11 * 1 KS763,00 KčSupplier Article Number: B153BKRXLLocal Article Number: PRTWB153BKRXL
Specifications
Garment applicationGeneral PurposeEnvironmentally PreferableRisk areaGenderFor menSleeve or leg lengthLongFabric93% wicking polyester, 7% elastane with silver ion treatmentWeight0,264 g/m² - PRTWB153BKRXXLB153BKRXXLBlackXXLRegular11 * 1 KS763,00 KčSupplier Article Number: B153BKRXXLLocal Article Number: PRTWB153BKRXXL
Specifications
Garment applicationGeneral PurposeEnvironmentally PreferableRisk areaGenderFor menSleeve or leg lengthLongFabric93% wicking polyester, 7% elastane with silver ion treatmentWeight0,284 g/m²