Safety underbench cabinets, type 90, UTS ergo line XL-5, 500 mm depth
Supplier: DUPERTHAL
Certificates
With classical wing door technology - 2 pull-out trays, 2 perforated sheet inserts and classical base 35 mm
Product Information
- Catalog #Supplier #DoorsW×D×HW×D×H int.WeightPkAvailabilityPrice
- Safety under bench cabinet UTS XL-5, carcass light grey RAL 7035, door zinc yellow RAL 1018Safety under bench cabinet UTS XL-5, carcass light grey RAL 7035, door zinc yellow RAL 1018DUEH29-061457-00529-061457-0053/classical1400×493×631 mm790×363×480
490×363×480 mm223 kg11 * 1 KS76 570,00 KčSupplier Article Number: 29-061457-005Local Article Number: DUEH29-061457-005Accessories
Specifications
Environmentally Preferable - Safety under bench cabinet UTS XL-5, carcass and door light grey RAL 7035Safety under bench cabinet UTS XL-5, carcass and door light grey RAL 7035DUEH29-061457-40529-061457-4053/classical1400×493×631 mm790×363×480
490×363×480 mm223 kg11 * 1 KS76 570,00 KčSupplier Article Number: 29-061457-405Local Article Number: DUEH29-061457-405Accessories
Specifications
Environmentally Preferable - Safety under bench cabinet UTS XL-5, carcass and door pure white RAL 9010Safety under bench cabinet UTS XL-5, carcass and door pure white RAL 9010DUEH29-061457-50529-061457-5053/classical1400×493×631 mm790×363×480
490×363×480 mm223 kg11 * 1 KS76 570,00 KčSupplier Article Number: 29-061457-505Local Article Number: DUEH29-061457-505Accessories
Specifications
Environmentally Preferable
With push-to-open wing door technology - 2 pull-out trays, 2 perforated sheet inserts and classical base 35 mm
Product Information
- Catalog #Supplier #DoorsW×D×HW×D×H int.WeightPkAvailabilityPrice
- Safety under bench cabinet UTS ST-XL-5, carcass and door light grey RAL 7035Safety under bench cabinet UTS ST-XL-5, carcass and door light grey RAL 7035DUEH29-061459-40529-061459-4053/push-to-open1400×493×681 mm790×363×480
490×363×480 mm223 kg11 * 1 KS80 230,00 KčSupplier Article Number: 29-061459-405Local Article Number: DUEH29-061459-405Accessories
Specifications
Environmentally Preferable - Safety under bench cabinet UTS XL-5, carcass light grey RAL 7035, door zinc yellow RAL 1018Safety under bench cabinet UTS XL-5, carcass light grey RAL 7035, door zinc yellow RAL 1018DUEH29-061459-00529-061459-0053/push-to-open1400×493×681 mm790×363×480
490×363×480 mm223 kg11 * 1 KS80 230,00 KčSupplier Article Number: 29-061459-005Local Article Number: DUEH29-061459-005Accessories