Tisk…

Decyl-β-D-glucopyranoside

Varování

MOLEM72640496EA 0 CZK
MOLEM72640496
Decyl-β-D-glucopyranoside
Decyl-β-D-glucopyranoside
Vzorec: C₁₆H₃₂O₆
MW: 320,43 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00063297
CAS číslo: 58846-77-8
EINECS: 261-469-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR