Tisk…

7-Chloroquinaldine

MOLEM66515008EA 0 CZK
MOLEM66515008
7-Chloroquinaldine
7-Chloroquinaldine
Vzorec: C₁₀H₈ClN
MW: 177,63 g/mol
MDL Number: MFCD00075436
CAS číslo: 4965-33-7
EINECS: 440-600-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR