Tisk…
Hygiene masks, PP
Dodavatel: GM Equipment
113-0687EA 813 CZK
113-0687
Hygiene masks, PP
Masky Roušky hygienické
This triple-thickness mask made from non woven, spunbonded PP provides complete safety and assures very comfortable breathing.
  • With integral nose clip
  • Three ply material, non-irritating
  • Microbiological filtration to 99,5%
Weight: 4 g (113-0687); 2 g (113-0688).