Tisk…
Surgical masks, THE LITE ONE*, PE/cellulose
Dodavatel: Kimberly-Clark
ANSM48105EA 386 CZK
ANSM48105
Surgical masks, THE LITE ONE*, PE/cellulose
The masks feature a flexible nosepiece for effective sealing and polyester cellulose inner layer to help prevent irritation.
  • Effectively filters 99,80% of particles at 0,1 µm
  • Effectively filters 97,84% of bacteria at 2,8 µm