Tisk…
Hledali jste: Sprchy na výplach očí
22  výsledků nalezeno
SearchResultCount:"22"
Sort Results
Zobrazit seznam Rychlý náhled (novinka)
Ohodnoťte výsledky hledání
Dodavatel: Plum Safety
Popis: Sterile sodium chloride solution (0,9%) in an eyewash bottle with eye cup. Helps prevent foreign bodies (such as metal and wood chips) from sticking t...
Katalogové číslo: (118-1701)
Dodavatel: Plum Safety
Popis: Wall-mounted station containing 1×500 ml eyewash complete with pictogram. Especially suitable for workplaces where on-the-spot rinsing is required.
Měrná jednotka: 1 * 1 KS
Dodavatel: Plum Safety
Popis: Sterile 0,9 % Sodium chloride solution is ideal for injuries with foreign bodies like dust, dirt ect. Sterile 4,9 % Phosphate buffer solution is perfe...
Dodavatel: Plum Safety
Popis: This wall station is equipped with two 1000 ml eyewash bottles, wall brackets, pictogram and mirror. The single bottle contains sterile sodium chlorid...
Dodavatel: Plum Safety
Popis: This wall-mountable dispenser contains 10 (2×5) ampoules with 20 ml of 0,9 % sterile sodium chloride eyewash solution.
Katalogové číslo: (118-0379)
Dodavatel: Plum Safety
Popis: This wall-mountable station is made of impact resistant plastic and equipped with a transparent cover. The station is specifically designed for use in...
Měrná jednotka: 1 * 1 KS
Katalogové číslo: (PLUM4862)
Dodavatel: Plum Safety
Popis: Wall-mounted eye wash box made from maintenance-free material for quick first aid of eye injuries. The box contains one 500 ml bottle of pH Neutral DU...
Měrná jednotka: 1 * 1 KS
Dodavatel: Plum Safety
Popis: Available as either an open station or closed and dust-proof wall box containing 2×500 ml plum eyewash bottles.
Dodavatel: Plum Safety
Popis: Wall-mounted eyewash stations, made in hard plastic, for simple and quick first-aid in case of an eye injury.
Dodavatel: Plum Safety
Popis: Sterile phosphate buffer solution (4,9%) for fast and effective neutralisation of acids and alkalis.
Katalogové číslo: (118-0477)
Dodavatel: Plum Safety
Popis: This wall-mountable dispenser contains 45 elastic plasters and 5 ampoules with 20 ml of 0,9% sterile sodium chloride eyewash solution. The elastic pla...
Měrná jednotka: 1 * 1 KS
Dodavatel: Plum Safety
Popis: Available as either an open station or closed, dust-proof wall box containing 200 ml pH neutral and 500 ml Plum eyewash. Ideal where there is a risk o...
Katalogové číslo: (118-0398)
Dodavatel: Plum Safety
Popis: This wall-mountable station is made of impact-resistant plastic and equipped with a transparent cover.
Měrná jednotka: 1 * 1 KS
Dodavatel: ORKLA CARE AB
Popis: Sterile, buffered, isotonic sodium chloride solution for rinsing eyes following an accident. Available as a single-use, pocket version (235 ml) or wal...
Katalogové číslo: (118-0277)
Dodavatel: ORKLA CARE AB
Popis: Wall-mountable eyewash station with two 500 ml bottles which open automatically when they are twisted out of their holders. The solution works to neut...
Měrná jednotka: 1 * 1 KS
Udržitelná balení
Udržitelné materiály
Katalogové číslo: (118-0253)
Dodavatel: ORKLA CARE AB
Popis: This plastic case with transparent lid and handle contains five bottles of Cederroth eyewash solution. The solution is intended for washing away parti...
Měrná jednotka: 1 * 1 KS
Cena na vyžádání
Omezené množství produktu na skladě. Zboží může být k dispozici v jiném skladě poblíž vašeho sídla. Přesvědčte se, že jste přihlášení na stránky, abyste mohli vidět dostupné položky na skladě. Pokud je stále zobrazeno call a potřebujete asistenci, volejte na číslo 321 570 321.
Omezené množství produktu na skladě. Zboží může být k dispozici v jiném skladě poblíž vašeho sídla. Přesvědčte se, že jste přihlášení na stránky, abyste mohli vidět dostupné položky na skladě. Pokud je stále zobrazeno call a potřebujete asistenci, volejte na číslo 321 570 321.
Daná chemikálie je regulována dle platné legislativy a bude vyžadováno vyplnění formuláře. Jeho včasným vyplněním uspíšíte dodání produktu.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Tento produkt je zablokován vaší organizací. Kontaktujte své nákupní oddělení pro více informací.
Původní produkt již není k dispozici. Zobrazen je náhradní produkt.
Tento produkt již není k dispozici. Podobné produkty můžete vyhledat pomocí VWR katalogových čísel nebo čísel dodavatele uvedených výše. Pokud potřebujete další asistenci, zavolejte na telefonní číslo zákaznického servisu VWR 321 570 321.
1 - 16 of 22
no targeter for Bottom