Erlenmeyer flasks, baffled, disposable, with screw cap, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
PETG, transparent, with HDPE screw cap.
- Leakproof HDPE screw cap closure can be opened partially for venting by twisting a quarter turn
- Packaged individually for easy storage and handling
- Reduced risk of cross-contamination
Sterile, disposable baffled Erlenmeyer flasks reduce the risk of cross-contamination. Ideal for shakers and suspension cultures, and for creating or storing media. Raised graduations for approximate reading of the contents.
Specifications
- Catalog No:
- 214-1928
- 214-1927
- 214-0142
- 214-0141
- 214-0140
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Specifications
Product Information
- Catalog #Supplier #CapacityThreadPkAvailabilityPrice
- 214-19284113-10001000 ml45-43061 * 6 KS3 130,00 KčSupplier Article Number: 4113-1000Local Article Number: NALG4113-1000Specifications:
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - 214-19274113-0125125 ml38-430241 * 24 KS8 030,00 KčSupplier Article Number: 4113-0125Local Article Number: NALG4113-0125Specifications:
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - 214-01424113-20002000 ml45-43041 * 4 KS3 310,00 KčSupplier Article Number: 4113-2000Local Article Number: NALG4113-2000Specifications:
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - 214-01414113-0500500 ml45-430121 * 12 KS5 540,00 KčSupplier Article Number: 4113-0500Local Article Number: NALG4113-0500Specifications:
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact - 214-01404113-0250250 ml38-430121 * 12 KS4 430,00 KčSupplier Article Number: 4113-0250Local Article Number: NALG4113-0250Specifications:
Low Manufacturing Impact Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact