Tisk…
Gas meters
Dodavatel: Ritter, part of Avantor
618-1133EA 0 CZK
RITATG3/6 RITATG5/6
Gas meters
These gas meters can measure volumes with no correction of temperature, humidity or gas type.
  • Maximum pressure 50 mbar
  • Hose barb connections
Precise measurement of the volume of gas flows, particularly suited to aggressive gases.
Informace o dodávce: Supplied with: four-chamber measuring drum, magnetic coupling, eight-digit roller counter, manometer/thermometer supports, viton seals, level and levelling feet.