Tisk…
Goggles, Vistamax 2000
Dodavatel: Honeywell Safety Products
111-3554EA 1030 CZK
DALL1002755
Goggles, Vistamax 2000
Designed for industrial, petrochemical or laboratory environments with mists, sprays or applications involving heat, humidity or physical exertion.
  • Adjustable headband in red
  • Extra deep frame profile
  • High flow ventilation and heat resistant
Fits comfortably over most prescription eyewear and with some half masks.
Weight: 142 g
Certifikace: EN 166, EN 170