Tisk…
Antibacterial lotion soap, Hy-G-Clenz™
Dodavatel: Micronova
Hy-G-Clenz™
129-0512EA 878 CZK
129-0512
Antibacterial lotion soap, Hy-G-Clenz™
Péče o pokožku Dezinfekce na ruce Kapalné a pěnové desinfekční prostředky na pleť
In 500 ml pump bottles, which can be used as stand alone or with VWR® touch free dispenser 129-0486. Hy-G-Clenz™ is an antibacterial soap lotion effective against a wide range of both gram positive and gram negative bacteria.
Hy-G-Clenz™ is an antibacterial soap lotion effective against a wide range of both gram positive and gram negative bacteria.
  • Unique blend of 0,3% Triclosan and natural moisturisers is ideal for frequent hand washing
  • Compliant with OSHA standards
  • Unique blend of 0,3% Triclosan and natural moisturisers is ideal for frequent hand washing
  • Compliant with OSHA standards
NovaClenz™ is a clear gel containing 60% ethyl alcohol and provides excellent antiseptic and sanitising capabilities. Helps to prevent transfer of bacterial agents, including pseudomonas, salmonella, and staphylococcus.
  • Instant hand sanitiser requires no sink, water or towels
  • Effective in reducing the incidence of nosocomial infections
AquaHol™ irradiated alcohol hand sanitiser is 70% gamma irradiated alcohol filtered to 0,1 µm.