Order Entry
Chamber slides, Nunc™ Lab-Tek™ II
undefined
Chamber slides, Nunc™ Lab-Tek™ II
  • Catalog No:
  • 734-2047
  • 734-2049
  • 734-2048
  • 734-2050
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  •  
    Recommended working volume
    Growth area
    Material
    Pk
  • One chamber
    One chamber
    734-2047
    154453
    2 – 4,5 ml
    8,6 cm²
    PS
    16
    1 * 16 KS
    1 * 16 KS - 6 350,00 Kč
    6 350,00 Kč
    1 * 16 KS
       6 350,00 Kč
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Two chambers
    Two chambers
    734-2048
    154461
    1 – 2 ml
    4 cm²
    PS
    16
    1 * 16 KS
    1 * 16 KS - 6 340,00 Kč
    6 340,00 Kč
    1 * 16 KS
       6 340,00 Kč
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Four chambers
    Four chambers
    734-2049
    154526
    0,5 – 1 ml
    1,7 cm²
    PS
    16
    1 * 16 KS
    1 * 16 KS - 6 220,00 Kč
    6 220,00 Kč
    1 * 16 KS
       6 220,00 Kč
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Eight chambers
    Eight chambers
    734-2050
    154534
    0,2 – 0,5 ml
    0,7 cm²
    PS
    16
    1 * 16 KS
    1 * 16 KS - 6 220,00 Kč
    6 220,00 Kč
    1 * 16 KS
       6 220,00 Kč
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact