Tisk…
Pipette tips, epT.I.P.S. Biopur® (IVD marked)
Dodavatel: Eppendorf
epT.I.P.S. Biopur®
613-3569EA 1600 CZK
613-3569 732-0095 613-3509 613-3570 613-3571 613-3510 613-3511
Pipette tips, epT.I.P.S. Biopur® (IVD marked)
Špičky pipetovací
These sterile tips are free of DNA, DNase, RNase and pyrogens. Conform to the highest standards for use in medical and pharmaceutical environments, the food industry, and in molecular and cellular biology applications.
Designed and constructed for low-contamination transfer of liquids, in particular for samples from the human body and for reagents within the scope of an in-vitro diagnostic application in order to allow the in-vitro diagnostic device to be used as intended. These epT.I.P.S. are in-vitro diagnostic devices according to Directive 98/79/EC of the European Parliament and the Council dated October 27, 1998.
Certifikace: With CE marking.
Balení: Available as epT.I.P.S.® Singles (packed individually) or as epT.I.P.S.® Racks (boxes containing racks of 96, 48 or 24 tips depending on tip volume).