Tisk…
VWR®, Stojánek pro práci s kryogenními vialkami
Dodavatel: VWR
VWR®
479-1869EA 1730 CZK
479-1869
VWR®, Stojánek pro práci s kryogenními vialkami
Stojany Stojánky na kryovialky
Constructed from sturdy polypropylene, the VWR® cryovial workstation rack is compact and stackable, with strong handles for safe and easy transport.
  • Holds up to 50 cryogenic vials
  • Autoclavable
  • Anti-skid rubber feet for bench top stability
  • Alphanumeric indexing for easy vial identification
Locking base allows for one hand cap assembly and removal with skirted cryovials.