Surgical scalpel handles and blades
About this item
Stainless steel.
- Slim and sleek with extra-fine blades for a precise cut
- Special scalpel handles for precise and easy handling
Blades are sterile and individually packed, handles are non sterile and individually packed.
Handles
Product Information
- Catalog #Supplier #TypePkAvailabilityPrice
- 233-00636051SF 111 * 1 KS1 800,00 KčSupplier Article Number: 6051Local Article Number: SWAN6051Specifications:
Accessories
Specifications
Environmentally Preferable - 233-00646052SF 211 * 1 KS1 800,00 KčSupplier Article Number: 6052Local Article Number: SWAN6052Specifications:
Accessories
Specifications
Environmentally Preferable - 233-00656053SF 311 * 1 KS1 800,00 KčSupplier Article Number: 6053Local Article Number: SWAN6053Specifications:
Accessories
Specifications
Environmentally Preferable - 233-00666054SF 411 * 1 KS1 800,00 KčSupplier Article Number: 6054Local Article Number: SWAN6054Specifications:
Accessories
Specifications
Environmentally Preferable - SWAN60626062SF 2311 * 1 KS2 180,00 KčSupplier Article Number: 6062Local Article Number: SWAN6062Specifications:
Accessories
Specifications
Environmentally Preferable
Blades
Product Information
- Catalog #SeqTypePkPriceAvailabilityPrice
- Supplier Article Number: 5904Local Article Number: SWAN5904