Tisk…
Hledali jste: Lopatky
108  výsledků nalezeno
SearchResultCount:"108"
Sort Results
Zobrazit seznam Rychlý náhled (novinka)
Ohodnoťte výsledky hledání
Dodavatel: VWR Collection
Popis: PS, bílá. Tyto sterilní odměrky jsou ideální pro odběr vzorků prášku, granulí a past.
Dodavatel: VWR Collection
Popis: The dual-purpose scoops are a unique economical solution that combines the function of a scoop and a weighing boat into one product. They are made fro...
Katalogové číslo: (231-0501)
Dodavatel: VWR Collection
Popis: PTFE, bílý.
Měrná jednotka: 1 * 1 KS
Dodavatel: RSG
Popis: Stainless steel. Manual scoops for collecting or measuring powders, pastes or granulates.
Dodavatel: BÜRKLE
Popis: Stainless steel V2A (1.4301), highly polished. Very stable hand scoop for various applications, particularly suitable for the food industry.
Dodavatel: BÜRKLE
Popis: PS, white. These scoops are ideal for the sampling of powder, granules and pastes. They are also available with a transparent lid to close it immediat...
Dodavatel: BÜRKLE
Popis: PS, white. These scoops are ideal for the sampling of powder, granules and pastes. They are also available with a transparent lid to close it immediat...
Dodavatel: BÜRKLE
Popis: The sampling scoop can be used to take samples of powders, granulates and pastes. The long handle makes it possible to sample larger containers even w...
Dodavatel: BÜRKLE
Popis: The ladle is suitable for taking samples of liquids and viscous media as well as powders and granulates. The ladle has a very smooth bottom and rounde...
Dodavatel: BÜRKLE
Popis: The SteriPlast® food scoop, made of blue PS, is produced and assembled in a class 7 cleanroom (10000, except for the 1000 ml scoop), individually pack...
Dodavatel: BÜRKLE
Popis: The sampling scoop can be used to take samples of powders, granulates and pastes. The long handle makes it possible to sample larger containers even w...
Udržitelné materiály
Dodavatel: BÜRKLE
Popis: The ladle is suitable for taking samples of liquids and viscous media as well as powders and granulates.
Dodavatel: BÜRKLE
Popis: These disposable scoops are manufactured from bio-plastic 'Green PE' which is entirely made of renewable resources. They are ideal for the sampling of...
Udržitelné materiály
Dodavatel: BÜRKLE
Popis: Universal graduated PP shovels for filling and measuring of powders and granules. For exact metering, the graduated shovel is to be overfilled and the...
Dodavatel: RSG
Popis: These chemical scoops are made from aluminium.
Dodavatel: BÜRKLE
Popis: The sampling scoop can be used to take samples of powders, granulates and pastes. The long handle makes it possible to sample larger containers even w...
Cena na vyžádání
Omezené množství produktu na skladě. Zboží může být k dispozici v jiném skladě poblíž vašeho sídla. Přesvědčte se, že jste přihlášení na stránky, abyste mohli vidět dostupné položky na skladě. Pokud je stále zobrazeno call a potřebujete asistenci, volejte na číslo 321 570 321.
Omezené množství produktu na skladě. Zboží může být k dispozici v jiném skladě poblíž vašeho sídla. Přesvědčte se, že jste přihlášení na stránky, abyste mohli vidět dostupné položky na skladě. Pokud je stále zobrazeno call a potřebujete asistenci, volejte na číslo 321 570 321.
Daná chemikálie je regulována dle platné legislativy a bude vyžadováno vyplnění formuláře. Jeho včasným vyplněním uspíšíte dodání produktu.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Tento produkt je zablokován vaší organizací. Kontaktujte své nákupní oddělení pro více informací.
Původní produkt již není k dispozici. Zobrazen je náhradní produkt.
Tento produkt již není k dispozici. Podobné produkty můžete vyhledat pomocí VWR katalogových čísel nebo čísel dodavatele uvedených výše. Pokud potřebujete další asistenci, zavolejte na telefonní číslo zákaznického servisu VWR 321 570 321.
1 - 16 of 108
no targeter for Bottom