Tisk…
Safety shoes, lace-up, 1600
Dodavatel: Abeba
114-2753EA 2160 CZK
ABEB1600/38
Safety shoes, lace-up, 1600
Leather shoes with leather lining and steel toecap.
  • Replaceable insole
  • Protective steel toecap (200 J)
  • PU sole with high slip resistance according to DIN 4843 part 100
Suitable for use in kitchens and in food processing industries.
Colour: White
Certifikace: EN ISO 20345; S2, SRC