Tisk…
Cell culture tubes, CELLSTAR®
Dodavatel: Greiner bio-one
CELLSTAR®
391-3405EA 9790 CZK
391-3405 391-3415 391-3410 391-3420
Cell culture tubes, CELLSTAR®
Zkumavky Zkumavky kultivační
PS, pre-treated, sterile.
  • Physical surface treatment for improved cell adhesion
  • Available with screw cap, bayonet cap or two position vented stopper
All tubes are certified free of RNase, DNase and human DNA, are non-pyrogenic and non-cytotoxic.