Tisk…
Centrifuge tubes, 13,5 ml, Sterilin®
Dodavatel: Thermo Fisher Scientific
Sterilin®
212-7401EA 9040 CZK
212-7401 525-0139 525-0135
Centrifuge tubes, 13,5 ml, Sterilin®
Zkumavky Zkumavky centrifugační
PS, graduated, round or conical bottom, non skirted, sterile or non sterile, with or without PE screw cap.
  • Moulded graduation marks at 5 and 10 ml
  • Wadded or non wadded screw cap
  • Recommended RCF 3200×g in suitably balanced rotors