Tisk…
Microcentrifuge tubes, 0,5/1,5/2,0 ml, Biopur® Safe-Lock, Eppendorf®
Dodavatel: Eppendorf
Biopur® Safe-Lock
211-2162EA 729 CZK
211-2162 211-2161 211-2160
Microcentrifuge tubes, 0,5/1,5/2,0 ml, Biopur® Safe-Lock, Eppendorf®
Zkumavky Mikrozkumavky centrifugační
PP, graduated, sterile, with attached cap, autoclavable when open.
  • Can withstand centrifugation up to 22,000xg for 0,5 ml, 1,5 ml, and 2,0 ml
  • The hook on the patented hinged lid clips around the rim of the test tube, preventing the tube from opening accidentally
  • For RNase-, DNA-, ATP- and pyrogen-free work
Balení: Individually sealed.