Order Entry
Large Dewar flasks, type C/CAL
Large Dewar flasks, type C/CAL
Supplier:  KGW
Certificates
undefined
Large Dewar flasks, type C/CAL
Supplier:  KGW
Specifications
  • Catalog No:
  • 478-4204
  • 478-0734
  • 478-4203
  • 478-4202
  • 478-4201
  • 478-0773
  • 478-0775
  • 478-0776
  • 478-0780
  • 478-0778
  • 478-0772
  • 478-0771
  • 478-0774
  • 478-0770
  • 478-0769
  • 478-4205
  • 478-0768
  • 478-0777
  • KGWA1252
  • Model:
  • 33 C
  • 32 C
  • 31 C
  • 30 C
  • 46
  • 30/7 C
  • 30 CAL
  • 30/4 CAL
  • 34 CAL
  • 44
  • 30/4 C
  • 34 C
  • 32 CAL
  • 31 CAL
  • Capacity:
  • 21 L
  • 14 L
  • 10 L
  • 7 L
  • 7 L
  • 7 L
  • 4 L
  • 28 L
  • 4 L
  • 28 L
  • 14 L
  • 10 L
  • Temperature range:
  • −200...+200 °C
  • −200...+200 °C
  • −200...+200 °C
  • −200...+200 °C
  • −200...+200 °C
  • −200...+200 °C
  • −200...+200 °C
  • −200...+200 °C
  • −200...+200 °C
  • −200...+200 °C
  • −200...+200 °C
  • −200...+200 °C
  • Daily evaporation:
  • 0,6 L
  • 0,4 L
  • 0,4 L
  • 0,45 L
  • 0,4 L
  • 0,45 L
  • 0,45 L
  • 0,6 L
  • 0,45 L
  • 0,6 L
  • 0,4 L
  • 0,4 L
  • Int.Ø:
  • 250 mm
  • 200 mm
  • 200 mm
  • 200 mm
  • 158 mm
  • 200 mm
  • 158 mm
  • 250 mm
  • 158 mm
  • 250 mm
  • 200 mm
  • 200 mm
  • Int. height:
  • 480 mm
  • 500 mm
  • 350 mm
  • 275 mm
  • 400 mm
  • 275 mm
  • 250 mm
  • 620 mm
  • 250 mm
  • 620 mm
  • 500 mm
  • 350 mm
  • Overall height:
  • 615 mm
  • 610 mm
  • 475 mm
  • 385 mm
  • 490 mm
  • 385 mm
  • 340 mm
  • 750 mm
  • 340 mm
  • 750 mm
  • 610 mm
  • 475 mm
Specifications
Type C: Metal - cover, blue coated
Product Information
  • Catalog #
    Supplier #
    For
    Pk
    Availability
    Price
  • Large Dewar flask, 33 C
    Large Dewar flask, 33 C
    478-4204
    1244
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 38 180,00 Kč
    38 180,00 Kč
    1 * 1 KS
       38 180,00 Kč
    Specifications:
    More
    Specifications
    Model
    33 C
    Capacity
    21 L
    Environmentally Preferable
    Temperature range
    −200...+200 °C
    Daily evaporation
    0,6 L
    Int.Ø
    250 mm
    Int. height
    480 mm
    Overall height
    615 mm
  • Large Dewar flask, 30 C
    Large Dewar flask, 30 C
    478-4201
    1241
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 18 040,00 Kč
    18 040,00 Kč
    1 * 1 KS
       18 040,00 Kč
    Specifications:
    More
    Specifications
    Model
    30 C
    Capacity
    7 L
    Environmentally Preferable
    Temperature range
    −200...+200 °C
    Daily evaporation
    0,45 L
    Int.Ø
    200 mm
    Int. height
    275 mm
    Overall height
    385 mm
  • Large Dewar flask, 31 C
    Large Dewar flask, 31 C
    478-4202
    1242
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 22 910,00 Kč
    22 910,00 Kč
    1 * 1 KS
       22 910,00 Kč
    Specifications:
    More
    Specifications
    Model
    31 C
    Capacity
    10 L
    Environmentally Preferable
    Temperature range
    −200...+200 °C
    Daily evaporation
    0,4 L
    Int.Ø
    200 mm
    Int. height
    350 mm
    Overall height
    475 mm
  • Large Dewar flask, 32 C
    Large Dewar flask, 32 C
    478-4203
    1243
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 23 570,00 Kč
    23 570,00 Kč
    1 * 1 KS
       23 570,00 Kč
    Specifications:
    More
    Specifications
    Model
    32 C
    Capacity
    14 L
    Environmentally Preferable
    Temperature range
    −200...+200 °C
    Daily evaporation
    0,4 L
    Int.Ø
    200 mm
    Int. height
    500 mm
    Overall height
    610 mm
  • Large Dewar flask, 34 C
    Large Dewar flask, 34 C
    478-4205
    1245
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 42 120,00 Kč
    42 120,00 Kč
    1 * 1 KS
       42 120,00 Kč
    Specifications:
    More
    Specifications
    Model
    34 C
    Capacity
    28 L
    Environmentally Preferable
    Temperature range
    −200...+200 °C
    Daily evaporation
    0,6 L
    Int.Ø
    250 mm
    Int. height
    620 mm
    Overall height
    750 mm
  • Large Dewar flask, 30/4 C
    Large Dewar flask, 30/4 C
    478-0774
    1248
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 16 180,00 Kč
    16 180,00 Kč
    1 * 1 KS
       16 180,00 Kč
    Specifications:
    More
    Specifications
    Model
    30/4 C
    Capacity
    4 L
    Environmentally Preferable
    Temperature range
    −200...+200 °C
    Daily evaporation
    0,45 L
    Int.Ø
    158 mm
    Int. height
    250 mm
    Overall height
    340 mm
  • Large Dewar flask, 30/7 C
    Large Dewar flask, 30/7 C
    478-0775
    1249
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 17 540,00 Kč
    17 540,00 Kč
    1 * 1 KS
       17 540,00 Kč
    Specifications:
    More
    Specifications
    Model
    30/7 C
    Capacity
    7 L
    Environmentally Preferable
    Temperature range
    −200...+200 °C
    Daily evaporation
    0,4 L
    Int.Ø
    158 mm
    Int. height
    400 mm
    Overall height
    490 mm
Type CAL: Stucco aluminium cover
Product Information
  • Catalog #
    Supplier #
    For
    Pk
    Availability
    Price
  • Large Dewar flask, 30 CAL
    Large Dewar flask, 30 CAL
    478-0776
    1251
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 18 040,00 Kč
    18 040,00 Kč
    1 * 1 KS
       18 040,00 Kč
    Specifications:
    More
    Specifications
    Model
    30 CAL
    Capacity
    7 L
    Environmentally Preferable
    Temperature range
    −200...+200 °C
    Daily evaporation
    0,45 L
    Int.Ø
    200 mm
    Int. height
    275 mm
    Overall height
    385 mm
  • Large Dewar flask, 31 CAL
    Large Dewar flask, 31 CAL
    KGWA1252
    1252
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 22 910,00 Kč
    22 910,00 Kč
    1 * 1 KS
       22 910,00 Kč
    Specifications:
    More
    Specifications
    Model
    31 CAL
    Capacity
    10 L
    Environmentally Preferable
    Temperature range
    −200...+200 °C
    Daily evaporation
    0,4 L
    Int.Ø
    200 mm
    Int. height
    350 mm
    Overall height
    475 mm
  • Large Dewar flask, 32 CAL
    Large Dewar flask, 32 CAL
    478-0777
    1253
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 23 570,00 Kč
    23 570,00 Kč
    1 * 1 KS
       23 570,00 Kč
    Specifications:
    More
    Specifications
    Model
    32 CAL
    Capacity
    14 L
    Environmentally Preferable
    Temperature range
    −200...+200 °C
    Daily evaporation
    0,4 L
    Int.Ø
    200 mm
    Int. height
    500 mm
    Overall height
    610 mm
  • Large Dewar flask, 34 CAL
    Large Dewar flask, 34 CAL
    478-0778
    1255
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 42 120,00 Kč
    42 120,00 Kč
    1 * 1 KS
       42 120,00 Kč
    Specifications:
    More
    Specifications
    Model
    34 CAL
    Capacity
    28 L
    Environmentally Preferable
    Temperature range
    −200...+200 °C
    Daily evaporation
    0,6 L
    Int.Ø
    250 mm
    Int. height
    620 mm
    Overall height
    750 mm
  • Large Dewar flask, 30/4 CAL
    Large Dewar flask, 30/4 CAL
    478-0780
    1258
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 16 180,00 Kč
    16 180,00 Kč
    1 * 1 KS
       16 180,00 Kč
    Specifications:
    More
    Specifications
    Model
    30/4 CAL
    Capacity
    4 L
    Environmentally Preferable
    Temperature range
    −200...+200 °C
    Daily evaporation
    0,45 L
    Int.Ø
    158 mm
    Int. height
    250 mm
    Overall height
    340 mm
Glass refill
Product Information
  • Catalog #
    Supplier #
    For
    Pk
    Availability
    Price
  • Glass refill, type 40
    Glass refill, type 40
    478-0734
    1231
    Large Dewar flask, 30/4 C and 30/4 CAL
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 9 330,00 Kč
    9 330,00 Kč
    1 * 1 KS
       9 330,00 Kč
    Specifications:
    Specifications
    Model
    Capacity
    Environmentally Preferable
    Temperature range
    Daily evaporation
    Int.Ø
    Int. height
    Overall height
  • Glass refill, type 41
    Glass refill, type 41
    478-0768
    1232
    Large Dewar flask, 30/7 C and 30/7 CAL
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 10 900,00 Kč
    10 900,00 Kč
    1 * 1 KS
       10 900,00 Kč
    Specifications:
    Specifications
    Model
    Capacity
    Environmentally Preferable
    Temperature range
    Daily evaporation
    Int.Ø
    Int. height
    Overall height
  • Glass refill, type 42
    Glass refill, type 42
    478-0769
    1233
    Large Dewar flask, 30 C and 30 CAL
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 11 290,00 Kč
    11 290,00 Kč
    1 * 1 KS
       11 290,00 Kč
    Specifications:
    Specifications
    Model
    Capacity
    Environmentally Preferable
    Temperature range
    Daily evaporation
    Int.Ø
    Int. height
    Overall height
  • Glass refill, type 43
    Glass refill, type 43
    478-0770
    1234
    Large Dewar flask, 31 C and 31 CAL
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 15 160,00 Kč
    15 160,00 Kč
    1 * 1 KS
       15 160,00 Kč
    Specifications:
    Specifications
    Model
    Capacity
    Environmentally Preferable
    Temperature range
    Daily evaporation
    Int.Ø
    Int. height
    Overall height
  • Glass refill, type 44
    Glass refill, type 44
    478-0771
    1235
    Large Dewar flask, 32 C and 32 CAL
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 16 470,00 Kč
    16 470,00 Kč
    1 * 1 KS
       16 470,00 Kč
    Specifications:
    More
    Specifications
    Model
    44
    Capacity
    Environmentally Preferable
    Temperature range
    Daily evaporation
    Int.Ø
    Int. height
    Overall height
  • Glass refill, type 46
    Glass refill, type 46
    478-0773
    1237
    Large Dewar flask, 34 C and 34 CAL
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 28 340,00 Kč
    28 340,00 Kč
    1 * 1 KS
       28 340,00 Kč
    Specifications:
    More
    Specifications
    Model
    46
    Capacity
    Environmentally Preferable
    Temperature range
    Daily evaporation
    Int.Ø
    Int. height
    Overall height
  • Glass refill, type 45
    Glass refill, type 45
    478-0772
    1236
    Large Dewar flask, 33 C and 33 CAL
    1
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 24 880,00 Kč
    24 880,00 Kč
    1 * 1 KS
       24 880,00 Kč
    Specifications:
    Specifications
    Model
    Capacity
    Environmentally Preferable
    Temperature range
    Daily evaporation
    Int.Ø
    Int. height
    Overall height