Tisk…
Hledali jste: Mineralizátory
26  výsledků nalezeno
SearchResultCount:"26"
Sort Results
Zobrazit seznam Rychlý náhled (novinka)
Ohodnoťte výsledky hledání
Dodavatel: Bohlender
Popis: Dimensionally stable basic vessel and screw cover made of TFM, homogenous, non porous surface.
Dodavatel: VELP SCIENTIFIC
Popis: A series of thermoreactors for digestion of samples for a range of analyses including COD, heavy metal, non metallic elements. The ECO 6 has the possi...
Dodavatel: VELP SCIENTIFIC
Popis: DK digesters are traditional digesters, consisting of an aluminium heating block that offers excellent thermal homogeneity, heating up to 450 °C, prov...
Dodavatel: VELP SCIENTIFIC
Popis: Fully automatic Kjeldahl digestion units for nitrogen analysis and protein determination that incorporate the TEMS™ technology for savings in terms of...
Katalogové číslo: (BEHRB00217639)
Dodavatel: BEHR
Popis: In the behrotest® nitrogen station, solvent is evaporated under ambient temperature conditions by using a mild nitrogen flow. A distributor leads the ...
Měrná jednotka: 1 * 1 KS
Katalogové číslo: (BEHRB00217638)
Dodavatel: BEHR
Popis: The complete sample clean up station includes dropping funnel, clean-up column with glass frit, reducing flasks and stand.
Měrná jednotka: 1 * 1 KS
Dodavatel: GESTIGKEIT HARRY
Popis: Rapid incinerator for individual and mass incineration of solid and liquid samples. Temperature range up to 950 °C.
Katalogové číslo: (705-0023)
Dodavatel: BEHR
Popis: The behrotest® universal analytical DET system is a flexible modular system designed for laboratories with frequently changing applications who do not...
Měrná jednotka: 1 * 1 KS
Katalogové číslo: (238-0122)
Dodavatel: GERHARDT
Popis: An automatic acid hydrolysis system for fat determination according to Weibull-Stoldt where the complete digestion and filtration process runs in a cl...
Měrná jednotka: 1 * 1 KS
Dodavatel: BEHR
Popis: Complete sets of equipment for preparation and distillation for commonly required analyses of solid and aqueous samples.
Katalogové číslo: (BUCH1154461000)
Dodavatel: BUCHI
Popis: The KjelDigester provides fast and safe digestion of up to 20 samples simultaneously.
Měrná jednotka: 1 * 1 KS
Dodavatel: BEHR
Popis: behrotest® InKjel digestion models are equipped with a high performance infrared heater.
Dodavatel: BUCHI
Popis: Digestion of protein samples with sulphuric acid and catalyst requires prolonged periods of heating and is traditionally carried out in block heaters....
Dodavatel: BEHR
Popis: COD determination is a very important parameter for wastewater and sewage measurements. Behr workstations guarantee COD analysis in strict accordance ...
Katalogové číslo: (BEHRB00217925)
Dodavatel: BEHR
Popis: The two-level behrosog process extraction system (neutraliser) – pre-separator plus safety level – keeps all acid vapours entirely away from the envir...
Měrná jednotka: 1 * 1 KS
Dodavatel: GERHARDT
Popis: Traditional Kjeldahl digestion units for flask sizes from 50 to 750 ml.
Cena na vyžádání
Omezené množství produktu na skladě. Zboží může být k dispozici v jiném skladě poblíž vašeho sídla. Přesvědčte se, že jste přihlášení na stránky, abyste mohli vidět dostupné položky na skladě. Pokud je stále zobrazeno call a potřebujete asistenci, volejte na číslo 321 570 321.
Omezené množství produktu na skladě. Zboží může být k dispozici v jiném skladě poblíž vašeho sídla. Přesvědčte se, že jste přihlášení na stránky, abyste mohli vidět dostupné položky na skladě. Pokud je stále zobrazeno call a potřebujete asistenci, volejte na číslo 321 570 321.
Daná chemikálie je regulována dle platné legislativy a bude vyžadováno vyplnění formuláře. Jeho včasným vyplněním uspíšíte dodání produktu.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Tento produkt je zablokován vaší organizací. Kontaktujte své nákupní oddělení pro více informací.
Původní produkt již není k dispozici. Zobrazen je náhradní produkt.
Tento produkt již není k dispozici. Podobné produkty můžete vyhledat pomocí VWR katalogových čísel nebo čísel dodavatele uvedených výše. Pokud potřebujete další asistenci, zavolejte na telefonní číslo zákaznického servisu VWR 321 570 321.
1 - 16 of 26
no targeter for Bottom