A fully automatic distillation system with integrated titration for precise analysis. The automated steam generator has steam output that can be set between 40 to 100%, and allows for all current procedures. In addition the system provides control of cooling water, sample tube and chemical tank as well as some safety features to protect the operator. The titration unit is fully integrated for the evaluation of the analysis using a potentiometric, automatic, end-point recognition.
- PC control and monitoring of process using VAPODEST® Manager software
- Online titration, during distillation is possible to optimise duration
- An Autosampler carousel is available and can be easily retrofitted for routine repetitive analyses
- Distillation time typically two to four minutes
- Nitrogen detection limit 0,1 mg
Other configurations of Autosampler carousel are available please ask Avantor specialists for further details.
Informace pro objednávání: VAPODEST® Software is included but not the control PC.