Tisk…
Cell disruptors, Sonifier®
Dodavatel: Branson
142-3703EA 117240 CZK
BRAN159-063-676 BRAN159-063-675
Cell disruptors, Sonifier®
These ultrasonic cell disruptors/homogenisers are versatile laboratory units suitable for a broad range of liquid processing applications such as biological cell or tissue disruption/homogenisation, extraction, emulsification, reaction acceleration, dispersion, fine mixing and degassing.
  • Analogue models have adjustable power output to meet each application's requirements and energy pulsing to minimise heating of sensitive samples
Informace o dodávce: Each model consists of a power supply, a converter and a mechanical probe or horn. The S-450A models have the choice of either a 12,7 mm or 19 mm horn, the larger horn is for use with maximum sample size of 500 ml. A large range of accessories is available, please order these separately.
Upozornení: Safety Note: Never use a probe with replacement tip with low surface tension liquids.