Tisk…
Melting point capillaries
Dodavatel: Cole Parmer
stuart®
621-2015EA 1670 CZK
621-2015 621-2016
Melting point capillaries
Zkumavky Kapiláry pro určení bodu tání
Manufactured from soda glass.
These tubes are easy to seal in a Bunsen flame and break into two.
Informace o dodávce: Supplied in robust tube holder.