Tisk…
Disc mill, DM 200
Dodavatel: Retsch
412-2101EA 606240 CZK
412-2101
Disc mill, DM 200
Mlýny
The DM 200 disc mill is used for batch-wise or continuous preliminary and fine size reduction of medium-hard to hard-brittle solids. The robust design, including a hinged grinding chamber for easy cleaning, makes the mill suitable for a wide range of materials and applications in laboratories and pilot plants.
  • Final fineness down to <100 µm
  • Short grinding times, typically a few minutes with inspection window for monitoring during operation
  • Accurate gap width setting between the grinding discs even during operation for reproducible results
  • Wide range of disc materials available for contaminant-free grinding
  • Connector for dust extraction
Informace pro objednávání: Grinding sets to be ordered separately.
Upozornení: * Depending on feed material.