Tisk…
Repetitive pipette, multi-channel, Finnpipette™ Multistepper
Dodavatel: Thermo Fisher Scientific
Finnpipette™
612-6301EA 0 CZK
612-6301 612-6300
Repetitive pipette, multi-channel, Finnpipette™ Multistepper
Pipety Pipety pro opakované použití
Lightweight and easy to use 8-channel micropipette, designed for dispensing liquids onto microplates and quick completion of routine laboratory tasks. It uses the same handpiece as the Finnpipette™ Stepper, and the 8-channels module is fitted to this.
  • Handpiece can be used with Finntip™ syringes (Finnpipette™ Stepper) or with an 8-channel module (Finnpipette™ Multistepper)
  • Eight channels are calibrated to dispense exactly the same volume of liquid at exactly the same time
  • Five pre-set volumes: 50, 100, 150, 200 and 250 µl
  • Dispenses with a single filling: 50 µl into 24×8 = 192 wells; 100 µl into 12×8 = 96 wells; 150 µl into 8×8 = 64 wells; 200 µl into 6×8 = 48 wells; 250 µl into 5×8 = 40 wells
  • Module rotates through 360° around the handpiece for increased comfort when pipetting
  • Complete volume chart is provided on the handpiece