Tisk…
Repetitive pipette, Varipette® 4720
Dodavatel: Eppendorf
Varipette®
612-7310EA 15710 CZK
612-7310
Repetitive pipette, Varipette® 4720
Pipety Pipety pro opakované použití
Pipette with continuously adjustable volume in increments of 10 µl, intended for accurate volumetric analysis up to 10 ml. It can be used for pipetting liquids with high vapour pressure or high viscosity.
  • Volume adjustable from 1 to 10 ml
  • Selector button for displaying the set point volume on a digital dial
  • Varitip P tips work on the principle of positive displacement, designed for pipetting volumes from 1 to 10 ml
  • Varitip S consists of a volumetric syringe and a Maxitip tip, works on the air displacement principle, and is designed for pipetting volumes from 2,5 to 10 ml into tall and narrow necked flasks
The Varipette® can be used with Varitip P or Varitip S tips for pipetting into different types of receptacles.