Tisk…
Flask shaker, SF1
Dodavatel: Cole Parmer
stuart®
444-0104EA 70040 CZK
444-0104
Flask shaker, SF1
Třepačky Třepačky rotační
Suitable for use with eight flasks up to 500 ml. Constructed from cast aluminium with metallic blue finish, and mounted on four vibration-absorbing rubber feet. Speed control is continuous using a control dial on the fascia panel. The side arms are secured to the central spindle by allen screws.
  • Built-in timer from 0 to 60 min or continuous mode, with manual override
  • Simulates hand shaking, ideal for extractions
  • Can be used at 5 to 40 °C, in an incubator or cold room
  • BioCote® antimicrobial protection
Certifikace: IP protection class according to DIN EN 60529: IP 20
Informace o dodávce: Supplied complete with eight clamps and Allen key.