Tisk…
Rockers, lab scale, digital, SSL3 and SSL4
Dodavatel: Cole Parmer
stuart®
444-0116EA 47330 CZK
444-0116 444-0115
Rockers, lab scale, digital, SSL3 and SSL4
Třepačky Třepačky rotační
Variable speed rockers with large platform are ideal for gentle mixing. Both models have an LED display with speed and time controlled via an easy to use encoder control knob.
  • Digital setting and display of speed and time
  • Can be used at 4 to 40 °C (80% relative humidity, non-condensing), in an incubator or cold room
  • Continuous or timer operation; integral digital timer 1 s to 90 min or from 1 min to 9 h, with automatic switch-off, time remaining is displayed
  • Optional, easy to fit, tier system triples the space available for samples
  • BioCote® antimicrobial protection
SSL3 3D gyratory rocker
The gentle 3D motion is ideal for foaming agitation, DNA extractions and staining or de-staining gels. The non-slip mat holds vessels in place and the optional tier system increases the working space.
SSL4 see-saw rocker
The see-saw (rocking) motion is ideal for washing and blotting procedures. The non-slip mat holds vessels in place, and the optional 2 tiers increases the working space.
Certifikace: IP protection class according to DIN EN 60529: IP 30
Informace o dodávce: Both units are supplied with non-slip mat, but without tier system.