Tisk…
Mechanical overhead stirrers, SS series
Dodavatel: Cole Parmer
Stuart®
441-0195EA 47010 CZK
441-0195 441-0208
Mechanical overhead stirrers, SS series
Míchadla Míchadla hřídelová
Range of quiet running overhead stirrers to meet all laboratory requirements.
  • SS20 has a hollow shaft which ensures easy adjustment of paddle height
  • BioCote® antimicrobial protection
  • Keyless chuck
SS10 is a general purpose stirrer, suitable for day to day laboratory use. With good speed control and overload protection. Ideal for aqueous and low viscosity liquids.
SS20 is a powerful stirrer for more demanding applications with advanced overload protection and sophisticated, responsive speed control. Suitable for low to medium viscosity liquids.
Certifikace: IP protection class according to DIN EN 60529: IP 42
Informace pro objednávání: Supplied without stand, paddle head or rod. Bossheads and stirring shafts must be ordered separately.