Overhead stirrer tools, anchor stirrers
Supplier: IKA
Certificates
About this item
Made from stainless steel. Ensures minimum deposits on vessel walls, and even distribution of high mineral content in liquids, ideal for mixing polymer reactions. High shearing forces at edges.
- Ideal for stirring liquids at low speeds
- Suitable for stirring liquids with medium to high viscosity
- Creates tangential flow
Product Information
- Katalogové čísloČíslo dodavateleStirrer ØShaft ØShaft lengthLengthMax. revolutionsPkAvailabilityPrice
- Anchor stirrerAnchor stirrer441-2931202230045 mm8 mm350 mm350 mm1000 min-11Log in for availability1 * 1 KS14 220,00 KčSpecifications:
Specifications
Environmentally Preferable - Anchor stirrerAnchor stirrer441-2932202240090 mm8 mm350 mm350 mm1000 min-11Log in for availability1 * 1 KS17 190,00 KčSpecifications:
Specifications
Environmentally Preferable - Anchor stirrerAnchor stirrer441-29342747400150 mm10 mm550 mm550 mm800 min-11Log in for availability1 * 1 KS27 980,00 KčSpecifications: