Tisk…
VWR®, Stopky digitální, vodotěsné
Dodavatel: VWR
VWR®
609-0131EA 3700 CZK
609-0131
VWR®, Stopky digitální, vodotěsné
Časovace/Stopky Stopky digitální
This electronic stopwatch includes a calendar and a clock.
  • Three-line display: time, countdown and pacer
  • Timing capacity: 10 hours
  • ¹/₁₀₀ second timing
  • Stores 30 lap times and calculates lap time average, allows for 99 split lap times
  • ¹⁰/₃₂₀ second pacer
Certifikace: Tested and calibrated in a triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST.
Informace o dodávce: Supplied with one CR2032 battery, certificate of calibration, individual serial number, and multi-language (English, French, German, Italian, Portuguese and Spanish) instructions.