Tisk…
Shaking water bath, THERMOLAB®
Dodavatel: GFL
462-5280EA 0 CZK
GFLA1070
Shaking water bath, THERMOLAB®
Versatile shaking water baths with reciprocating motion. Bath has four separate stainless steel compartments, in which different samples can be heated independently of each other. Alternatively different users can use the same bath at the same time for different temperature-dependent reactions. Ideal for PCR related tasks.
  • Four separate compartments with cover for each
  • Separate microprocessor controlled temperature regulation and display for each compartment
  • Common shaking action for all compartments
  • Lids are double walled and angled inside, which prevents condensation dripping into samples
  • Height adjustable sample holders can be used with or without optional racks. Microtitre plates, Western Blot vessels, centrifuge tubes, ampoules or bottles can be fitted into the individual compartments
Informace pro objednávání: Supplied without racks.