(-)-Quinine
Dodavatel: EHRENSTORFER GMBH
EHERC16706900EA
1750
CZK
EHERC16706900
(-)-Quinine
(-)-Quinine
Vzorec:
C₂₀H₂₄N₂O₂ MW: 324,42 g/mol Bod tání: 176 °C |
MDL Number:
MFCD00198096 CAS číslo: 130-95-0 EINECS: 205-003-2 Merck Index: 12,08245 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...