Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Czech Republic
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0,00 Kč
Order Entry
Home
Flasks
Cell Culture Flasks
Carboxyl cell culture flasks, Corning® PureCoat™
Carboxyl cell culture flasks, Corning® PureCoat™
Carboxyl cell culture flasks, Corning® PureCoat™
734-2408
:
Corning
Carboxyl cell culture flasks, Corning® PureCoat™
734-2408
:
Corning
: 356780
: BDAA356780
12 330,00 Kč
1 * 40 KS
-
+
Closure type:
Vented cap
Growth area:
175 cm²
Packed:
Pack of 5, Case of 40
Capacity:
750 ml
Seq:
04
Neck:
Straight
Pk:
40