Tisk…
Endothelial cell growth environment, Corning® BioCoat™
Dodavatel: Corning Life Sciences
734-1082EA 11600 CZK
BDAA355053
Endothelial cell growth environment, Corning® BioCoat™
Corning® BioCoat™ Endothelial Cell Growth Environment is an integrated system designed to create in vitro intestinal models. The system comprises low serum medium, culture supplements, growth factors, and Corning® BioCoat™ Collagen I Cultureware in one convenient kit.