


Acetaldehyde ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75-07-0
:
Formula:CH₃CHO
MDL Number:MFCD00006991
:44.05 g/mol
Merck Index:14,39
Boiling Pt:20.8 °C
Melting Pt:–128 °C
Density:0.780
Flash Pt:–27 °C (–17 °F)
Storage Temperature:Ambient
EINECS:200-836-8
UN:1089
ADR:3,I
:GHS02
:GHS08
:GHS07