Tisk…

Furan-2-boronic acid

Varování

MOLEM84472096EA 0 CZK
MOLEM84472096
Furan-2-boronic acid
Furan-2-boronic acid
Vzorec: C₄H₅BO₃
MW: 111,89 g/mol
MDL Number: MFCD00799544
CAS číslo: 13331-23-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR