3-Bromo-5-(trifluoromethyl)benzenesulphonyl chloride, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
SIAL558613-1GEA
2770
CZK
SIAL558613-1G
3-Bromo-5-(trifluoromethyl)benzenesulphonyl chloride, Sigma-Aldrich®
3-Bromo-5-(trifluoromethyl)benzenesulphonyl chloride
Vzorec:
C₇H₃BrClF₃O₂S MW: 323,52 g/mol Bod varu: 217…218 °C (1013 hPa) Hustota: 1,835 g/cm³ (20 °C) |
MDL Number:
MFCD03094393 CAS číslo: 351003-46-8 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...