Pentacosanoic acid
Dodavatel: EHRENSTORFER GMBH
EHERC15973630EA
6200
CZK
EHERC15973630
Pentacosanoic acid
Pentacosanoic acid
Vzorec:
C₂₅H₅₀O₂ MW: 382,67 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00020551 CAS číslo: 506-38-7 EINECS: 208-036-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...