Tisk…

n-Tetradekan ≥99%

Dodavatel: ABCR GMBH
Nebezpečí

ABCRAB118948.0500EA 0 CZK
ABCRAB118948.0500
n-Tetradekan ≥99%
n-Tetradekan

Purity: 99%
CAS No.: 629-59-4
Molecular Formula: C14H30
Formula Weight: 198.39
Flash Point: 99°(211°F)
Physical Form: Liquid
Density: 0.763
Refractive Index: 1.4290
Appearance: Colourless
Odour: Mild
Melting Point: 6°
Boiling Point: 252-254°
MDL No.: MFCD00008986

Vzorec: H₃C(CH₂)₁₂CH₃
MW: 198,39 g/mol
Bod varu: 252…254 °C
Bod tání: 6 °C
Hustota: 0,763
Bod vzplanutí: 99 °C (211 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008986
CAS číslo: 629-59-4
EINECS: 211-096-0
https://cz.vwr.com/stibo/web/std.lang.all/86/19/738619.jpg

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR