Tisk…
Ultrafiltration cassettes, Sartocon®
Dodavatel: Sartorius
Sartocon®
512-0066EA 12030 CZK
512-0066
Ultrafiltration cassettes, Sartocon®
Filtry Tangenciální kazety
Hydrosart® membranes are stabilised cellulose based membranes that have been optimised for the biotechnological and pharmaceutical industry. It is a stable polymer that features a broad pH range. It is also extremely hydrophilic, making it non-protein binding. virtually non-foul and has extremely high flux. Membrane regeneration, storage and depyrogenation can be accomplished by using NaOH even at elevated temperatures.
  • Filtration area: 0,02 m²
  • Bacterial endotoxins: <0,18 EU/ml as determined by the LAL test
  • pH stability: pH 2 to 14
Membranes are designed for use in the biotechnological and pharmaceutical industries. They can be used for the following applications: oligonucleotide, proteins (albumin - even with 40% EtOH - hemoglobin), coagulation factors (factor VIII factor III), vaccines (tetanus, diphteria), monoclonal antibodies.