Methylakrylát
Dodavatel: EHRENSTORFER GMBH
EHERCA10045400EA
1600
CZK
EHERCA10045400
Methylakrylát
Methylakrylát
Vzorec:
C₄H₆O₂ MW: 86,09 g/mol |
MDL Number:
MFCD00008627 CAS číslo: 96-33-3 EINECS: 202-500-6 UN: 1919 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...