Tisk…

5-Methoxysalicylic acid

MOLE54516788-25GEA 0 CZK
MOLE54516788-25G
5-Methoxysalicylic acid
5-Methoxysalicylic acid
Vzorec: C₈H₈O₄
MW: 168,15 g/mol
Bod varu: 327 °C (1013 hPa)
Bod tání: 141…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00002459
CAS číslo: 2612-02-4
EINECS: 220-037-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR