Tisk…
Ductless filter fume hoods, Chemcap™ Pro2 series
Dodavatel: Bigneat
481-0637EA 254910 CZK
BIGNPR2008/OS BIGNPR2008/MP BIGNPR2008/MP/HP BIGNPR1004/OS/HP BIGNPR1004/AM/HP BIGNPR1506/MP/HP BIGNPR1506/OS/HP BIGNPR1004/MP BIGNPR2008/OS/HP BIGNPR1004/HPLS/HP BIGNPR1004/MP/HP BIGNPR1004/HPLUS BIGNPR1004/OS BIGNPR1506/MP BIGNPR1004/F/HP BIGNPR1506/OS BIGNPR1004/F BIGNPR1004/AM
Ductless filter fume hoods, Chemcap™ Pro2 series
The Chemcap™ Pro2 cabinets (ductless, recirculatory filtration) provide increased operator safety over the Chemcap™ Clearview™ cabinet range, achieved by double carbon filtration and HEPA particulate filtration and powerful extraction system. Chemcap™ Pro2 is designed for use with the most toxic fumes, vapours and particulates. This cabinet incorporates a back-up filter – when the lower filter reaches saturation point, any fumes breaking through are retained by the upper back-up filter. To facilitate periodic testing of the interfilter gases, the filter housing contains an integral sampling chamber. During service the main filter is removed and the back-up filter is dropped down to become the main filter and a new back-up filter installed.
Cabinets are manufactured from epoxy coated steel and transparent panels are of clear acrylic and are highly resistant to corrosion.
  • Excellent all-round vision
  • Keypad control panel with touch button controls and service due indicator
  • Audible electronic low air flow alarm
  • Cabinets include lighting and trapezium front access as standard with double hinged and lockable front door flaps for easy access to cabinet interior
  • The PVC base tray offers excellent chemical resistance and contains spillages
Cabinet can be placed onto an existing work surface or it may be supplied with mobile bench or static stand. Select either a mobile bench with integrated cupboard or an open box section bench (castors/fixed height). A range of three standard cabinet sizes.
Operator protection: Double filtration protection, double safe.
Chemcap carbon/HEPA filtration options: Filters are included - most frequently used MP (multipurpose) and OS (where organic fumes predominate) are listed below, with or without HP (HP refers to HEPA filtration). Alternative filter types H+ (acids), F (formaldehyde), AM (ammonia) are available on request.
Power supply: 230 V, AC, 50 Hz, 5 Amp, 1 Ø (on request 110 V, AC, 60 Hz, 8 Amp, 1 Ø).
Certifikace: Chemcap™ Pro2 conforms to the requirements of all major standards for containment and filtration:
- US Standard ASHRAE 110: 1995 12-203 (for containment)
- British Standard BS 7989:2001 (for filter efficiency and retention capacity)
- French Standard AFNOR NFX 15-211:2009 - Class 2 (for filter retention capacity)
Informace pro objednávání: Please contact us for details on alternative filter types and other options, including electronic filter saturation alarm, mobile stands and service options.
Informace o dodávce: Cabinets are supplied with a full set of filters (pre-filter, 2x carbon filters of pre-determined type, and with or without HEPA filter), a rear baffle and lighting (18 W fluorescent lamps). Please note that the cabinets are delivered unassembled.