2,6-Dimethoxybenzoic acid
MOLE13772930-100GEA
0
CZK
MOLE13772930-100G
2,6-Dimethoxybenzoic acid
2,6-Dimethoxybenzoic acid
Vzorec:
C₉H₁₀O₄ MW: 182,18 g/mol Bod varu: 315 °C (1013 hPa) Bod tání: 180…182 °C Hustota: 1,365 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002437 CAS číslo: 1466-76-8 EINECS: 215-985-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...